Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGGTGGCCCCGGAGGACGTAGGC[C/G]TTTCCGGCCCCTCGCGACCGAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603371 MIM: 602015 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLE1 - GLE1, RNA export mediator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003722.1 | Intron | NP_001003722.1 | ||||
NM_001499.2 | Intron | NP_001490.1 | ||||
XM_006717059.2 | Intron | XP_006717122.1 | ||||
XM_006717060.2 | Intron | XP_006717123.1 | ||||
XM_011518549.1 | Intron | XP_011516851.1 | ||||
XM_011518550.1 | Intron | XP_011516852.1 | ||||
XM_011518551.1 | Intron | XP_011516853.1 |
ODF2 - outer dense fiber of sperm tails 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |