Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 169730 MIM: 611115 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGA5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PGA5 - pepsinogen 5, group I (pepsinogen A) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWCE - von Willebrand factor C and EGF domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152718.2 | 2401 | Silent Mutation | CGA,CGG | R,R 843 | NP_689931.2 | |
XM_006718460.1 | 2401 | Silent Mutation | CGA,CGG | R,R 835 | XP_006718523.1 | |
XM_006718462.1 | 2401 | Silent Mutation | CGA,CGG | R,R 783 | XP_006718525.1 | |
XM_011544817.1 | 2401 | Silent Mutation | CGA,CGG | R,R 745 | XP_011543119.1 | |
XM_017017340.1 | 2401 | Silent Mutation | CGA,CGG | R,R 822 | XP_016872829.1 | |
XM_017017341.1 | 2401 | Silent Mutation | CGA,CGG | R,R 800 | XP_016872830.1 | |
XM_017017342.1 | 2401 | Silent Mutation | CGA,CGG | R,R 713 | XP_016872831.1 | |
XM_017017343.1 | 2401 | Silent Mutation | CGA,CGG | R,R 560 | XP_016872832.1 | |
XM_017017344.1 | 2401 | Intron | XP_016872833.1 |