Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATACCTGTCACTCCAAAGGACTCT[C/T]CTCAGCAGCCAGGTAGGATGAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609244 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAET1E-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAET1E-AS1 - RAET1E antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAET1G - retinoic acid early transcript 1G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001788.3 | 964 | Silent Mutation | AGA,AGG | R,R 274 | NP_001001788.2 | |
XM_011535800.2 | 964 | Silent Mutation | AGA,AGG | R,R 274 | XP_011534102.1 | |
XM_017010831.1 | 964 | Silent Mutation | AGA,AGG | R,R 274 | XP_016866320.1 |
Set Membership: |
HapMap |