Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAATTTATAACAATTCCGTTTCA[C/T]CCAAATGGTAAGAACTAAATGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2U PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2U - ubiquitin conjugating enzyme E2 U (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152489.1 | 1213 | Silent Mutation | CAC,CAT | H,H 78 | NP_689702.1 | |
XM_005270498.4 | 1213 | Silent Mutation | CAC,CAT | H,H 78 | XP_005270555.1 | |
XM_006710379.3 | 1213 | Silent Mutation | CAC,CAT | H,H 46 | XP_006710442.1 | |
XM_011540764.2 | 1213 | Silent Mutation | CAC,CAT | H,H 78 | XP_011539066.1 | |
XM_017000379.1 | 1213 | Intron | XP_016855868.1 | |||
XM_017000380.1 | 1213 | Intron | XP_016855869.1 | |||
XM_017000381.1 | 1213 | Intron | XP_016855870.1 | |||
XM_017000382.1 | 1213 | Intron | XP_016855871.1 | |||
XM_017000383.1 | 1213 | Intron | XP_016855872.1 | |||
XM_017000384.1 | 1213 | Intron | XP_016855873.1 | |||
XM_017000385.1 | 1213 | Intron | XP_016855874.1 |