Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGACATAACACAAATCTTCCACA[C/T]AAAGGCAGAAATACAGTATCAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606469 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM55 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM55 - tripartite motif containing 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033058.2 | Intron | NP_149047.2 | ||||
NM_184085.1 | Intron | NP_908973.1 | ||||
NM_184086.1 | Intron | NP_908974.1 | ||||
NM_184087.1 | Intron | NP_908975.1 | ||||
XM_005251316.4 | Intron | XP_005251373.1 | ||||
XM_011517613.2 | Intron | XP_011515915.1 | ||||
XM_011517614.2 | Intron | XP_011515916.1 | ||||
XM_011517615.2 | Intron | XP_011515917.1 | ||||
XM_011517616.2 | Intron | XP_011515918.1 | ||||
XM_011517617.2 | Intron | XP_011515919.1 | ||||
XM_017013908.1 | Intron | XP_016869397.1 | ||||
XM_017013909.1 | Intron | XP_016869398.1 |