Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTTTGAGTGAATCCTACAAATA[C/G]AGTGAATGAGATAAAACTAACATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300170 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OFD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OFD1 - OFD1, centriole and centriolar satellite protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003611.2 | Intron | NP_003602.1 | ||||
XM_005274604.2 | Intron | XP_005274661.1 | ||||
XM_005274606.3 | Intron | XP_005274663.1 | ||||
XM_011545592.2 | Intron | XP_011543894.1 | ||||
XM_011545594.2 | Intron | XP_011543896.1 | ||||
XM_011545595.1 | Intron | XP_011543897.1 | ||||
XM_011545597.1 | Intron | XP_011543899.1 | ||||
XM_011545598.1 | Intron | XP_011543900.1 | ||||
XM_017029909.1 | Intron | XP_016885398.1 | ||||
XM_017029910.1 | Intron | XP_016885399.1 | ||||
XM_017029911.1 | Intron | XP_016885400.1 |