Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAACGCTTATTTTTTGACCGGA[C/T]ATTCTGGCTATACCGCATCATCATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDAD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SDAD1 - SDA1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288983.1 | 2155 | Intron | NP_001275912.1 | |||
NM_001288984.1 | 2155 | Intron | NP_001275913.1 | |||
NM_018115.3 | 2155 | Intron | NP_060585.2 | |||
XM_005263102.4 | 2155 | Missense Mutation | ATC,GTC | I,V 635 | XP_005263159.1 | |
XM_005263105.4 | 2155 | Intron | XP_005263162.1 | |||
XM_017008354.1 | 2155 | Intron | XP_016863843.1 | |||
XM_017008355.1 | 2155 | Intron | XP_016863844.1 |