Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCTGAGTTCCAGGACAGCCTC[A/C]ACTTCAGCTACCATCCCTCAGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 157680 MIM: 609372 MIM: 609373 | ||||||||||||||||||||
Literature Links: |
CDC25C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC25C - cell division cycle 25C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM53C - family with sequence similarity 53 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135647.1 | 641 | Missense Mutation | AAC,CAC | N,H 66 | NP_001129119.1 | |
NM_016605.2 | 641 | Missense Mutation | AAC,CAC | N,H 66 | NP_057689.1 |
KDM3B - lysine demethylase 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |