Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTTTTTTTTTTTTTTTTTAAGG[C/T]TTGTAAACATATAACATAAAAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602533 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PARK7 - Parkinsonism associated deglycase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123377.1 | Intron | NP_001116849.1 | ||||
NM_007262.4 | Intron | NP_009193.2 | ||||
XM_005263424.3 | Intron | XP_005263481.1 |