Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAGTCAGCAGATTTCTGTCTGG[C/T]TAGAGAAAAACATCTTACCCTGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF800 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF800 - zinc finger protein 800 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176814.4 | Intron | NP_789784.2 | ||||
XM_005250181.3 | Intron | XP_005250238.1 | ||||
XM_006715867.3 | Intron | XP_006715930.1 | ||||
XM_006715868.3 | Intron | XP_006715931.1 | ||||
XM_006715869.3 | Intron | XP_006715932.1 | ||||
XM_011515859.2 | Intron | XP_011514161.1 |