Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGCCAAGTCTGGACAATTCTC[A/C]CAGTCCCTTGAACTCCTCGAGGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHNAK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHNAK2 - AHNAK nucleoprotein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C14orf79 - chromosome 14 open reading frame 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174891.3 | 438 | Silent Mutation | TCA,TCC | S,S 95 | NP_777551.2 | |
XM_005267317.3 | 438 | Silent Mutation | TCA,TCC | S,S 95 | XP_005267374.1 | |
XM_005267318.3 | 438 | Silent Mutation | TCA,TCC | S,S 95 | XP_005267375.1 |