Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTCTGTTCTCAACAAAGGCGTA[G/A]GTTGGGAGCGACTGAGCCACTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609650 MIM: 617032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP6 - NLR family pyrin domain containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGGHG - protein-glucosylgalactosylhydroxylysine glucosidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025092.4 | 180 | Intron | NP_079368.3 | |||
XM_011520383.2 | 180 | UTR 5 | XP_011518685.1 | |||
XM_011520384.2 | 180 | UTR 5 | XP_011518686.1 | |||
XM_011520385.2 | 180 | UTR 5 | XP_011518687.1 | |||
XM_011520386.2 | 180 | UTR 5 | XP_011518688.1 | |||
XM_011520387.2 | 180 | UTR 5 | XP_011518689.1 | |||
XM_017018355.1 | 180 | Intron | XP_016873844.1 |