Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATGTAAAATTATACTTTTCATG[C/T]CTACGTTGTATGAACTTAATTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605489 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT81 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFT81 - intraflagellar transport 81 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143779.1 | Intron | NP_001137251.1 | ||||
NM_014055.3 | Intron | NP_054774.2 | ||||
NM_031473.3 | Intron | NP_113661.2 | ||||
XM_011538232.2 | Intron | XP_011536534.1 | ||||
XM_017019217.1 | Intron | XP_016874706.1 | ||||
XM_017019218.1 | Intron | XP_016874707.1 |