Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGCCGCTTGGCGCCTGGCCCA[C/T]GCTTGTGCCTCCCCCTGCAGCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611179 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107984115 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107984115 - laforin-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHROOM1 - shroom family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172700.1 | 2117 | Silent Mutation | NP_001166171.1 | |||
NM_133456.2 | 2117 | Silent Mutation | NP_597713.2 | |||
XM_005271885.4 | 2117 | Silent Mutation | XP_005271942.1 | |||
XM_005271886.3 | 2117 | Silent Mutation | XP_005271943.1 | |||
XM_006714534.3 | 2117 | Intron | XP_006714597.1 | |||
XM_011543167.2 | 2117 | Silent Mutation | XP_011541469.1 |
SOWAHA - sosondowah ankyrin repeat domain family member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |