Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGATACATAATTACAGCAAGC[A/C]CTGCTTAGGAACTTTGCACGCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610216 MIM: 608536 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO8 - anoctamin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020959.2 | Intron | NP_066010.1 | ||||
XM_017027048.1 | Intron | XP_016882537.1 |
GTPBP3 - GTP binding protein 3 (mitochondrial) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128855.2 | Intron | NP_001122327.1 | ||||
NM_001195422.1 | Intron | NP_001182351.1 | ||||
NM_032620.3 | Intron | NP_116009.2 | ||||
NM_133644.3 | Intron | NP_598399.2 |