Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTGGCACCTATCACAAGGCTT[G/T]ATCTACAAGGAGGCAATCTATTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602929 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VWA5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VWA5A - von Willebrand factor A domain containing 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130142.1 | Intron | NP_001123614.1 | ||||
NM_014622.4 | Intron | NP_055437.2 | ||||
NM_198315.2 | Intron | NP_938057.1 | ||||
XM_011542828.2 | Intron | XP_011541130.1 |