Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTGAGAAGTCCCTTTCTGAGCT[A/G]TCATAGCCCTGAGATAAGTCATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610528 MIM: 605012 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHD8 - chromodomain helicase DNA binding protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170629.1 | 6975 | Silent Mutation | GAC,GAT | D,D 2555 | NP_001164100.1 | |
NM_020920.3 | 6975 | Silent Mutation | GAC,GAT | D,D 2276 | NP_065971.2 |
SNORD9 - small nucleolar RNA, C/D box 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUPT16H - SPT16 homolog, facilitates chromatin remodeling subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |