Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613927 MIM: 138470 MIM: 154040 MIM: 600478 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2 - complement component 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CFB - complement factor B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1236 - microRNA 1236 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NELFE - negative elongation factor complex member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002904.5 | Intron | NP_002895.3 | ||||
XM_006715205.3 | Intron | XP_006715268.1 | ||||
XM_011514913.2 | Intron | XP_011513215.1 | ||||
XM_017011299.1 | Intron | XP_016866788.1 |
SKIV2L - Ski2 like RNA helicase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |