Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCCCTGCAGTCATCTCGGGGG[A/C]TCCTGAGGGTCCAGGTCCACATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603392 MIM: 614479 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BIK - BCL2 interacting killer | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCAT - malonyl-CoA-acyl carrier protein transacylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014507.3 | 1210 | UTR 3 | NP_055322.1 | |||
NM_173467.4 | 1210 | Silent Mutation | NP_775738.3 | |||
XM_011530156.2 | 1210 | Missense Mutation | XP_011528458.1 |