Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAACTGCCTCCCACTACCGTATT[A/C]ACAATATGATTAACTCGTGTTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611315 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZSCAN20 - zinc finger and SCAN domain containing 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145238.3 | Intron | NP_660281.2 | ||||
XM_005271171.3 | Intron | XP_005271228.1 | ||||
XM_006710874.3 | Intron | XP_006710937.1 | ||||
XM_006710876.3 | Intron | XP_006710939.1 | ||||
XM_017002237.1 | Intron | XP_016857726.1 | ||||
XM_017002238.1 | Intron | XP_016857727.1 | ||||
XM_017002239.1 | Intron | XP_016857728.1 | ||||
XM_017002240.1 | Intron | XP_016857729.1 | ||||
XM_017002241.1 | Intron | XP_016857730.1 |
Set Membership: |
HapMap |