Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGGAAAGAGAGGCGTAGTCTT[C/T]GCTTCATTCTTTTTGAAGACCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAN1B1 - mannosidase alpha class 1B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016219.4 | Intron | NP_057303.2 | ||||
XM_006716945.3 | Intron | XP_006717008.1 | ||||
XM_017014239.1 | Intron | XP_016869728.1 |
MAN1B1-AS1 - MAN1B1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UAP1L1 - UDP-N-acetylglucosamine pyrophosphorylase 1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |