Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCGGGTCCTCTGAGGTGAAACC[C/G]CAAAGGAAAGCGGGATCGGGACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606818 MIM: 609827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPP3 - dipeptidyl peptidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256670.1 | 63 | UTR 5 | NP_001243599.1 | |||
NM_005700.4 | 63 | UTR 5 | NP_005691.2 | |||
NM_130443.3 | 63 | UTR 5 | NP_569710.2 |
LOC101928069 - uncharacterized LOC101928069 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PELI3 - pellino E3 ubiquitin protein ligase family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |