Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAACTTGTACTTTTTGCCAAGT[C/G]TTCAACTCTTCATTGAGCTATCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300255 | ||||||||||||||||||||
Literature Links: |
OGT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OGT - O-linked N-acetylglucosamine (GlcNAc) transferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181672.2 | 760 | Intron | NP_858058.1 | |||
NM_181673.2 | 760 | Intron | NP_858059.1 | |||
XM_017029907.1 | 760 | Intron | XP_016885396.1 | |||
XM_017029908.1 | 760 | UTR 5 | XP_016885397.1 |