Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTACTGGCCTGGCCCTCAGCCCA[A/G]ACACCCAGGCTCACTTGGTGAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609906 MIM: 611461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFS - embryonal Fyn-associated substrate | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A17 - solute carrier family 22 member 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289050.1 | Intron | NP_001275979.1 | ||||
NM_016609.4 | Intron | NP_057693.3 | ||||
NM_020372.3 | Intron | NP_065105.2 | ||||
XM_005267747.4 | Intron | XP_005267804.1 | ||||
XM_005267748.4 | Intron | XP_005267805.1 | ||||
XM_017021361.1 | Intron | XP_016876850.1 | ||||
XM_017021362.1 | Intron | XP_016876851.1 |