Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTGCCACTGGCCAAGGCCAAGA[T/C]CTGGGCGTTGGGGGAGAGGGCCAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605842 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TBL2 - transducin (beta)-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012453.2 | 1037 | Missense Mutation | ATC,GTC | I,V 345 | NP_036585.1 | |
XM_006715923.3 | 1037 | Missense Mutation | ATC,GTC | I,V 312 | XP_006715986.1 | |
XM_011516027.2 | 1037 | Missense Mutation | ATC,GTC | I,V 180 | XP_011514329.1 |