Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCGTCTTTAAAGGGCCTCCTAC[A/G]AGCTACTCTTGGGCCATATGGGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138319 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHURC1-FNTB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHURC1-FNTB - CHURC1-FNTB readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202558.1 | 2317 | Intron | NP_001189487.1 | |||
NM_001202559.1 | 2317 | Intron | NP_001189488.1 |
GPX2 - glutathione peroxidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB15 - RAB15, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308154.1 | 2317 | UTR 3 | NP_001295083.1 | |||
NM_198686.2 | 2317 | UTR 3 | NP_941959.1 | |||
XM_017021277.1 | 2317 | UTR 3 | XP_016876766.1 |
Set Membership: |
HapMap |