Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCACTTCACTGAGAAAGTCTAT[C/G]GTTTTCAGATTTCCATGCCATTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAIP1 - poly(A) binding protein interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006451.4 | Intron | NP_006442.2 | ||||
NM_182789.3 | Intron | NP_877590.1 | ||||
NM_183323.2 | Intron | NP_899152.1 | ||||
XM_005248230.3 | Intron | XP_005248287.1 | ||||
XM_017008956.1 | Intron | XP_016864445.1 |