Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610162 MIM: 608676 | ||||||||||||||||||||
Literature Links: |
CCDC28B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC28B - coiled-coil domain containing 28B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301011.1 | Intron | NP_001287940.1 | ||||
NM_024296.4 | Intron | NP_077272.2 | ||||
XM_011542113.2 | Intron | XP_011540415.1 | ||||
XM_017002306.1 | Intron | XP_016857795.1 | ||||
XM_017002307.1 | Intron | XP_016857796.1 | ||||
XM_017002308.1 | Intron | XP_016857797.1 | ||||
XM_017002309.1 | Intron | XP_016857798.1 |
DCDC2B - doublecortin domain containing 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IQCC - IQ motif containing C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM234 - transmembrane protein 234 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019118.4 | Intron | NP_061991.3 | ||||
XM_011541795.2 | Intron | XP_011540097.1 | ||||
XM_017001812.1 | Intron | XP_016857301.1 | ||||
XM_017001813.1 | Intron | XP_016857302.1 | ||||
XM_017001814.1 | Intron | XP_016857303.1 | ||||
XM_017001815.1 | Intron | XP_016857304.1 | ||||
XM_017001816.1 | Intron | XP_016857305.1 | ||||
XM_017001817.1 | Intron | XP_016857306.1 | ||||
XM_017001818.1 | Intron | XP_016857307.1 | ||||
XM_017001819.1 | Intron | XP_016857308.1 | ||||
XM_017001820.1 | Intron | XP_016857309.1 | ||||
XM_017001821.1 | Intron | XP_016857310.1 | ||||
XM_017001822.1 | Intron | XP_016857311.1 | ||||
XM_017001823.1 | Intron | XP_016857312.1 | ||||
XM_017001824.1 | Intron | XP_016857313.1 | ||||
XM_017001825.1 | Intron | XP_016857314.1 | ||||
XM_017001826.1 | Intron | XP_016857315.1 | ||||
XM_017001827.1 | Intron | XP_016857316.1 | ||||
XM_017001828.1 | Intron | XP_016857317.1 | ||||
XM_017001829.1 | Intron | XP_016857318.1 | ||||
XM_017001830.1 | Intron | XP_016857319.1 | ||||
XM_017001831.1 | Intron | XP_016857320.1 |
TXLNA - taxilin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |