Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCAAATATACATTCTTGTTCCC[A/G]AAAACATCCCAGGCAGTTTCAGCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608713 MIM: 602047 | ||||||||||||||||||||
Literature Links: |
CYP2R1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP2R1 - cytochrome P450 family 2 subfamily R member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024514.4 | 1830 | UTR 3 | NP_078790.2 | |||
XM_005252788.1 | 1830 | UTR 3 | XP_005252845.1 | |||
XM_005252789.2 | 1830 | UTR 3 | XP_005252846.1 | |||
XM_011519895.1 | 1830 | UTR 3 | XP_011518197.1 | |||
XM_011519898.2 | 1830 | UTR 3 | XP_011518200.1 | |||
XM_017017190.1 | 1830 | UTR 3 | XP_016872679.1 | |||
XM_017017191.1 | 1830 | UTR 3 | XP_016872680.1 | |||
XM_017017192.1 | 1830 | UTR 3 | XP_016872681.1 | |||
XM_017017193.1 | 1830 | UTR 3 | XP_016872682.1 | |||
XM_017017194.1 | 1830 | UTR 3 | XP_016872683.1 | |||
XM_017017195.1 | 1830 | UTR 3 | XP_016872684.1 |
PDE3B - phosphodiesterase 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |