Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTTTACGTTCCACCTGCGTCTC[A/G]GAAATGTCAGACCCTGGAACTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606505 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723313 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC102723313 - uncharacterized LOC102723313 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PINX1 - PIN2/TERF1 interacting, telomerase inhibitor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |