Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATTATCTACCTCTATGTGCTA[C/T]AAGATGGTCTTCAACTTCTACTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616210 MIM: 231675 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf46 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C4orf46 - chromosome 4 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ETFDH - electron transfer flavoprotein dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281737.1 | 448 | Intron | NP_001268666.1 | |||
NM_001281738.1 | 448 | Intron | NP_001268667.1 | |||
NM_004453.3 | 448 | Missense Mutation | ACA,ATA | T,I 31 | NP_004444.2 |
Set Membership: |
HapMap |