Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCGATGCACTAACATTTAACCTGT[C/T]CCTGGTGACTTCTGAATCACTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182888 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZP2 - zona pellucida glycoprotein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290104.1 | Intron | NP_001277033.1 | ||||
NM_003460.2 | Intron | NP_003451.1 | ||||
XM_011545942.2 | Intron | XP_011544244.1 |