Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCAGAGAATCCTGAGACAACCC[A/G]GAGGAGCCCCTGGGGGCAGCAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRDM15 - PR domain 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040424.2 | 4640 | UTR 3 | NP_001035514.1 | |||
NM_001282934.1 | 4640 | UTR 3 | NP_001269863.1 | |||
NM_022115.4 | 4640 | Intron | NP_071398.3 | |||
XM_006724039.2 | 4640 | UTR 3 | XP_006724102.1 | |||
XM_006724040.2 | 4640 | UTR 3 | XP_006724103.1 | |||
XM_011529674.1 | 4640 | UTR 3 | XP_011527976.1 | |||
XM_011529675.1 | 4640 | UTR 3 | XP_011527977.1 | |||
XM_011529676.2 | 4640 | UTR 3 | XP_011527978.1 | |||
XM_011529677.2 | 4640 | UTR 3 | XP_011527979.1 | |||
XM_011529678.2 | 4640 | UTR 3 | XP_011527980.1 | |||
XM_011529679.2 | 4640 | UTR 3 | XP_011527981.1 | |||
XM_011529681.2 | 4640 | UTR 3 | XP_011527983.1 | |||
XM_011529683.1 | 4640 | UTR 3 | XP_011527985.1 | |||
XM_017028425.1 | 4640 | UTR 3 | XP_016883914.1 | |||
XM_017028426.1 | 4640 | UTR 3 | XP_016883915.1 |