Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTCACTAATGGATGGTCTTGT[A/G]TGACCAAGTGCAGTCATTAAATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614518 MIM: 602136 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GATAD1 - GATA zinc finger domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021167.4 | 3851 | Intron | NP_066990.3 |
PEX1 - peroxisomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000466.2 | 3851 | Silent Mutation | CAC,CAT | H,H 1240 | NP_000457.1 | |
NM_001282677.1 | 3851 | Silent Mutation | CAC,CAT | H,H 1183 | NP_001269606.1 | |
NM_001282678.1 | 3851 | Silent Mutation | CAC,CAT | H,H 1032 | NP_001269607.1 | |
XM_017012319.1 | 3851 | Silent Mutation | CAC,CAT | H,H 657 | XP_016867808.1 |
Set Membership: |
HapMap |