Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCTTGCTGGGTGCTGGTCTTGG[A/C]AGGGCCATCTACTGGGGATAGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608016 MIM: 611853 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COA4 - cytochrome c oxidase assembly factor 4 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016565.2 | 720 | UTR 3 | NP_057649.2 | |||
XM_017017883.1 | 720 | UTR 3 | XP_016873372.1 | |||
XM_017017884.1 | 720 | UTR 3 | XP_016873373.1 |
MRPL48 - mitochondrial ribosomal protein L48 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAAF1 - proteasomal ATPase associated factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |