Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCGGCCGTGGACTCCTATCCCA[A/G]CTGGCTCAAGTTCCACATTGGTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611061 | ||||||||||||||||||||
Literature Links: |
FAM20C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM20C - family with sequence similarity 20 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020223.3 | 915 | Missense Mutation | AAC,AGC | N,S 225 | NP_064608.2 | |
XM_017012450.1 | 915 | Missense Mutation | AAC,AGC | N,S 225 | XP_016867939.1 | |
XM_017012451.1 | 915 | Missense Mutation | AAC,AGC | N,S 225 | XP_016867940.1 | |
XM_017012452.1 | 915 | Missense Mutation | AAC,AGC | N,S 225 | XP_016867941.1 | |
XM_017012453.1 | 915 | Missense Mutation | AAC,AGC | N,S 225 | XP_016867942.1 | |
XM_017012454.1 | 915 | Missense Mutation | AAC,AGC | N,S 225 | XP_016867943.1 | |
XM_017012455.1 | 915 | Intron | XP_016867944.1 |