Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCATCTTGAATGTGACAAAGGCTC[A/G]CATAGCTGCTGGCTTACCAATGGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606444 MIM: 612988 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREBZF PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CREBZF - CREB/ATF bZIP transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM126A - transmembrane protein 126A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244735.1 | 276 | UTR 5 | NP_001231664.1 | |||
NM_032273.3 | 276 | Intron | NP_115649.1 |