Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGAGAGTCCTCGAACAAAGAAA[A/T]TTCCACTAACTGAAGAGGAAATATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605494 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EFCAB7 - EF-hand calcium binding domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032437.3 | 329 | Missense Mutation | ATT,TTT | I,F 27 | NP_115813.2 | |
XM_005271274.3 | 329 | Missense Mutation | ATT,TTT | I,F 27 | XP_005271331.1 | |
XM_006710976.3 | 329 | Missense Mutation | ATT,TTT | I,F 40 | XP_006711039.1 | |
XM_006710977.1 | 329 | Missense Mutation | ATT,TTT | I,F 27 | XP_006711040.1 | |
XM_011542301.2 | 329 | Missense Mutation | ATT,TTT | I,F 40 | XP_011540603.1 | |
XM_017002548.1 | 329 | Missense Mutation | ATT,TTT | I,F 40 | XP_016858037.1 |
ITGB3BP - integrin subunit beta 3 binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |