Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTTAGGAATGGAGTAATTGTTC[A/G]TGTAGGCAATGGTGTCATCCATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNGB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNGB3 - cyclic nucleotide gated channel beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019098.4 | 1445 | Missense Mutation | ACG,ATG | T,M 466 | NP_061971.3 | |
XM_011517138.2 | 1445 | Missense Mutation | ACG,ATG | T,M 328 | XP_011515440.1 |