Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAGAAGGACAAGTCTGTCTAGG[A/G]GCAGTAATGTCCACAGGACGTAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPC - heterogeneous nuclear ribonucleoprotein C (C1/C2) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077442.1 | Intron | NP_001070910.1 | ||||
NM_001077443.1 | Intron | NP_001070911.1 | ||||
NM_004500.3 | Intron | NP_004491.2 | ||||
NM_031314.2 | Intron | NP_112604.2 | ||||
XM_006720125.2 | Intron | XP_006720188.1 | ||||
XM_011536708.1 | Intron | XP_011535010.1 | ||||
XM_011536709.2 | Intron | XP_011535011.1 | ||||
XM_011536710.2 | Intron | XP_011535012.1 | ||||
XM_011536711.2 | Intron | XP_011535013.1 | ||||
XM_011536712.1 | Intron | XP_011535014.1 | ||||
XM_017021252.1 | Intron | XP_016876741.1 | ||||
XM_017021253.1 | Intron | XP_016876742.1 |
LINC00641 - long intergenic non-protein coding RNA 641 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |