Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCATTATTTACCTAATGAAATTG[G/T]GAAAAACTATACATATTCAGCAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602545 | ||||||||||||||||||||
Literature Links: |
PTPRK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPRK - protein tyrosine phosphatase, receptor type K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135648.2 | 5559 | UTR 3 | NP_001129120.1 | |||
NM_001291981.1 | 5559 | UTR 3 | NP_001278910.1 | |||
NM_001291982.1 | 5559 | Intron | NP_001278911.1 | |||
NM_001291983.1 | 5559 | Intron | NP_001278912.1 | |||
NM_001291984.1 | 5559 | UTR 3 | NP_001278913.1 | |||
NM_002844.3 | 5559 | UTR 3 | NP_002835.2 |