Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCGGCCGATCCTTGCGGAGGTG[C/T]GACTGGCCCGGATCATCTCTGTCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 238310 MIM: 611516 MIM: 600690 | |||||||||||||||||||||||
Literature Links: |
AMT PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AMT - aminomethyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NICN1 - nicolin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032316.3 | 657 | Missense Mutation | CAC,CGC | H,R 191 | NP_115692.1 |
TCTA - T-cell leukemia translocation altered | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |