Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCCTGCATGAGCACTAGTTGT[C/T]GTCATTATTATTTATCATAATTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFSWAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SFSWAP - splicing factor SWAP homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261411.1 | Intron | NP_001248340.1 | ||||
NM_004592.3 | Intron | NP_004583.2 | ||||
XM_011538653.1 | Intron | XP_011536955.1 | ||||
XM_011538654.2 | Intron | XP_011536956.1 | ||||
XM_011538655.2 | Intron | XP_011536957.1 | ||||
XM_017019798.1 | Intron | XP_016875287.1 | ||||
XM_017019799.1 | Intron | XP_016875288.1 |