Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATGCACTCTCCTTTTGACCCCTA[C/T]AATTCACCCTATGCCCAAATCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC30A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC30A3 - solute carrier family 30 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318949.1 | Intron | NP_001305878.1 | ||||
NM_001318950.1 | Intron | NP_001305879.1 | ||||
NM_001318951.1 | Intron | NP_001305880.1 | ||||
NM_003459.4 | Intron | NP_003450.2 | ||||
XM_005264547.1 | Intron | XP_005264604.1 | ||||
XM_005264548.3 | Intron | XP_005264605.1 | ||||
XM_006712100.2 | Intron | XP_006712163.1 | ||||
XM_011533102.2 | Intron | XP_011531404.1 | ||||
XM_011533103.2 | Intron | XP_011531405.1 | ||||
XM_017004873.1 | Intron | XP_016860362.1 | ||||
XM_017004874.1 | Intron | XP_016860363.1 | ||||
XM_017004875.1 | Intron | XP_016860364.1 |