Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGTTTGCTACCAAACCTGGAGG[C/T]GGGGCTTTGGTTTTGCTTGAGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137028 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM227B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALK2 - galactokinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001556.2 | 2091 | Silent Mutation | GGC,GGT | G,G 439 | NP_001001556.1 | |
NM_001289030.1 | 2091 | Silent Mutation | GGC,GGT | G,G 426 | NP_001275959.1 | |
NM_001289031.1 | 2091 | Silent Mutation | GGC,GGT | G,G 426 | NP_001275960.1 | |
NM_002044.3 | 2091 | Silent Mutation | GGC,GGT | G,G 450 | NP_002035.1 | |
XM_005254279.3 | 2091 | Intron | XP_005254336.1 | |||
XM_005254280.3 | 2091 | Intron | XP_005254337.1 | |||
XM_005254284.3 | 2091 | Intron | XP_005254341.1 | |||
XM_006720461.3 | 2091 | Intron | XP_006720524.1 | |||
XM_006720462.3 | 2091 | Intron | XP_006720525.1 | |||
XM_006720463.3 | 2091 | Intron | XP_006720526.1 | |||
XM_006720464.3 | 2091 | Intron | XP_006720527.1 | |||
XM_011521441.2 | 2091 | Intron | XP_011519743.1 | |||
XM_017022062.1 | 2091 | Silent Mutation | GGC,GGT | G,G 393 | XP_016877551.1 | |
XM_017022063.1 | 2091 | Intron | XP_016877552.1 | |||
XM_017022064.1 | 2091 | Intron | XP_016877553.1 | |||
XM_017022065.1 | 2091 | Intron | XP_016877554.1 | |||
XM_017022066.1 | 2091 | Intron | XP_016877555.1 |