Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610530 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4638 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR4638 - microRNA 4638 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM41 - tripartite motif containing 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033549.4 | 967 | Missense Mutation | ACT,GCT | T,A 78 | NP_291027.3 | |
NM_201627.2 | 967 | Missense Mutation | ACT,GCT | T,A 78 | NP_963921.1 | |
XM_006714929.2 | 967 | Missense Mutation | ACT,GCT | T,A 78 | XP_006714992.1 | |
XM_006714930.2 | 967 | Missense Mutation | ACT,GCT | T,A 78 | XP_006714993.1 | |
XM_006714931.3 | 967 | Missense Mutation | ACT,GCT | T,A 78 | XP_006714994.1 | |
XM_006714932.3 | 967 | Missense Mutation | ACT,GCT | T,A 78 | XP_006714995.1 |
Set Membership: |
HapMap |