Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCATCACCAACCAAGTCTCCTC[A/T]CGGAAGGTAGTGCTATCTGCTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM81A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM81A - family with sequence similarity 81 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152450.2 | 286 | Intron | NP_689663.2 | |||
XM_005254166.2 | 286 | Intron | XP_005254223.1 | |||
XM_006720398.3 | 286 | UTR 5 | XP_006720461.1 | |||
XM_006720399.2 | 286 | Intron | XP_006720462.1 | |||
XM_011521247.2 | 286 | UTR 5 | XP_011519549.1 | |||
XM_011521248.2 | 286 | Intron | XP_011519550.1 | |||
XM_011521249.2 | 286 | UTR 5 | XP_011519551.1 | |||
XM_011521250.2 | 286 | Intron | XP_011519552.1 | |||
XM_017021931.1 | 286 | UTR 5 | XP_016877420.1 | |||
XM_017021932.1 | 286 | Intron | XP_016877421.1 |