Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615136 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK2A3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CSNK2A3 - casein kinase 2 alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256686.1 | Intron | NP_001243615.1 |
GALNT18 - polypeptide N-acetylgalactosaminyltransferase 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198516.2 | Intron | NP_940918.2 | ||||
XM_006718224.2 | Intron | XP_006718287.1 | ||||
XM_006718225.2 | Intron | XP_006718288.1 | ||||
XM_011520068.2 | Intron | XP_011518370.1 | ||||
XM_011520069.2 | Intron | XP_011518371.1 | ||||
XM_011520070.2 | Intron | XP_011518372.1 | ||||
XM_011520071.2 | Intron | XP_011518373.1 |
Set Membership: |
HapMap |