Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGGGCCAAGGTTCCAGCATCC[A/G]CAGCCATCAGCCATGTCCCTGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600022 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGIR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTGIR - prostaglandin I2 (prostacyclin) receptor (IP) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000960.3 | 2084 | UTR 3 | NP_000951.1 | |||
XM_005259093.2 | 2084 | UTR 3 | XP_005259150.1 | |||
XM_005259095.3 | 2084 | UTR 3 | XP_005259152.1 |