Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGAAGCACAGGGGCAAAACCCC[C/G]CACCCCAAATCCTGCAACCCAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600509 MIM: 600937 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCC8 - ATP binding cassette subfamily C member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000352.4 | Intron | NP_000343.2 | ||||
NM_001287174.1 | Intron | NP_001274103.1 | ||||
XM_011520331.2 | Intron | XP_011518633.1 | ||||
XM_017018197.1 | Intron | XP_016873686.1 | ||||
XM_017018198.1 | Intron | XP_016873687.1 | ||||
XM_017018199.1 | Intron | XP_016873688.1 | ||||
XM_017018200.1 | Intron | XP_016873689.1 | ||||
XM_017018201.1 | Intron | XP_016873690.1 | ||||
XM_017018202.1 | Intron | XP_016873691.1 | ||||
XM_017018203.1 | Intron | XP_016873692.1 | ||||
XM_017018204.1 | Intron | XP_016873693.1 |
KCNJ11 - potassium voltage-gated channel subfamily J member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |